Allylcyclohexan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL662844-5GEA
84.9
EUR
SIAL662844-5G
SIAL662844-25G
Allylcyclohexan, Sigma-Aldrich®
Allylcyclohexan
Formel:
C₉H₁₆ Molecular Weight: 124,23 g/mol |
MDL:
MFCD00013769 CAS-Nummer: 2114-42-3 EINECS: 218-316-4 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...