1,2:5,6-Diepoxycyclooctan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL160466-10GEA 134 EUR
SIAL160466-10G
1,2:5,6-Diepoxycyclooctan, Sigma-Aldrich®
1,2:5,6-Diepoxycyclooctane
Formel: C₈H₁₂O₂
MDL: MFCD00005076
CAS-Nummer: 27035-39-8
EINECS: 248-180-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR