1,2:5,6-Diepoxycyclooctan

Lieferant: CHEMPUR

CHMPFL13520.POREA 0 EUR
CHMPFL13520.POR
1,2:5,6-Diepoxycyclooctan
1,2:5,6-Diepoxycyclooctane
Formel: C₈H₁₂O₂
MDL: MFCD00005076
CAS-Nummer: 27035-39-8
EINECS: 248-180-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR