cis-Bicyclo[3.3.0]octan-3,7-dion, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL192104-1GEA 154 EUR
SIAL192104-1G
cis-Bicyclo[3.3.0]octan-3,7-dion, Sigma-Aldrich®
cis-Bicyclo[3.3.0]octan-3,7-dion
Formel: C₈H₁₀O₂
Molecular Weight: 138,1600 g/mol
EINECS: 257-357-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR