2,4-Dimethoxy-N-(4-fluorobenzyl)aniline, 97%

H57556.03EA 153 EUR
ALFAH57556.03 ALFAH57556.MD
2,4-Dimethoxy-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019594-96-7
Molecular Formula: C15H16FNO2
Formula Weight: 261.29
MDL No.: MFCD11924930
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR