N-(4-Fluorobenzyl)-3,4-dimethoxyaniline, 97%

H57820.MDEA 47.8 EUR
ALFAH57820.MD ALFAH57820.03
N-(4-Fluorobenzyl)-3,4-dimethoxyaniline, 97%
Purity: 97%
CAS No.: 1019542-69-8
Molecular Formula: C15H16FNO2
Formula Weight: 261.29
MDL No.: MFCD11924932
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR