3,5-Difluoro-N-(2-fluorobenzyl)aniline, 97%

H56955.MDEA 38.3 EUR
ALFAH56955.MD ALFAH56955.03
3,5-Difluoro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1021023-11-9
Molecular Formula: C13H10F3N
Formula Weight: 237.22
MDL No.: MFCD11146637
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR