3-Chloro-N-(2-fluorobenzyl)-4-methoxyaniline, 97%

H57757.MDEA 27.3 EUR
ALFAH57757.MD ALFAH57757.03
3-Chloro-N-(2-fluorobenzyl)-4-methoxyaniline, 97%
Purity: 97%
CAS No.: 940362-92-5
Molecular Formula: C14H13ClFNO
Formula Weight: 265.71
MDL No.: MFCD11058265
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR