4-Chloro-2-fluoro-N-(2-fluorobenzyl)aniline, 97%

H57101.MDEA 38.3 EUR
ALFAH57101.MD ALFAH57101.03
4-Chloro-2-fluoro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1152634-83-7
Molecular Formula: C13H10ClF2N
Formula Weight: 253.67
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR