3,4-Dichloro-N-(3-fluorobenzyl)aniline, 97%

H57135.03EA 170 EUR
ALFAH57135.03 ALFAH57135.MD
3,4-Dichloro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1036555-59-5
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD11116346
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR