3,4-Dichloro-N-(2-fluorobenzyl)aniline, 97%

H57544.03EA 85 EUR
ALFAH57544.03 ALFAH57544.MD
3,4-Dichloro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 723753-73-9
Molecular Formula: C13H10Cl2FN
Formula Weight: 270.13
MDL No.: MFCD05740290
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR