2-Bromo-4-fluoro-N-(4-fluorobenzyl)aniline, 97%

H57108.03EA 139 EUR
ALFAH57108.03 ALFAH57108.MD
2-Bromo-4-fluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1039960-80-9
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD11189242
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR