4-Bromo-2-fluoro-N-(3-fluorobenzyl)aniline, 97%

H57203.03EA 191 EUR
ALFAH57203.03 ALFAH57203.MD
4-Bromo-2-fluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019562-83-4
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD11142753
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR