4-Bromo-2-chloro-N-(4-fluorobenzyl)aniline, 97%

H57109.03EA 127 EUR
ALFAH57109.03 ALFAH57109.MD
4-Bromo-2-chloro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1097814-35-1
Molecular Formula: C13H10BrClF
Formula Weight: 300.58
MDL No.: MFCD11643666
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR