2,6-Bis(thiocyanatomethyl)-4-methylanisol

Lieferant: CHEMPUR

XXXXB2332.5EA 0 EUR
XXXXB2332.5 CHMPB2332.11 XXXXB2332.1
2,6-Bis(thiocyanatomethyl)-4-methylanisol
2,6-Bis(thiocyanatomethyl)-4-methylanisol
Formel: C₁₂H₁₂N₂OS₂
CAS-Nummer: 206559-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR