2,5-Bis(brommethyl)-4-(2-ethylhexyloxy)anisol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL536539-1GEA
306
EUR
SIAL536539-1G
2,5-Bis(brommethyl)-4-(2-ethylhexyloxy)anisol, Sigma-Aldrich®
2,5-Bis(brommethyl)-4-(2-ethylhexyloxy)anisol
Formel:
C₁₇H₂₆Br₂O₂ |
MDL:
MFCD03093944 CAS-Nummer: 209625-37-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...