3,5-Dimethyl-4-n-propoxybenzeneboronic acid, 98%

H53294.03EA 31.5 EUR
ALFAH53294.03 ALFAH53294.06
3,5-Dimethyl-4-n-propoxybenzeneboronic acid, 98%
Purity: 98%
CAS No.: 357611-51-9
Molecular Formula: C11H17BO3
Formula Weight: 208.06
Melting Point: 170-173°
MDL No.: MFCD05865178
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR