3-Chloro-N-(3-fluorobenzyl)-2-methylaniline, 97%

H57955.03EA 255 EUR
ALFAH57955.03 ALFAH57955.MD
3-Chloro-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1036531-56-2
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
MDL No.: MFCD11115760
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR