4-Bromo-N-(4-fluorobenzyl)-3-methylaniline, 97%

H57347.03EA 69.6 EUR
ALFAH57347.03 ALFAH57347.MD
4-Bromo-N-(4-fluorobenzyl)-3-methylaniline, 97%
Purity: 97%
CAS No.: 1019487-46-7
Molecular Formula: C14H13BrFN
Formula Weight: 294.16
MDL No.: MFCD11142808
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR