4-(2-Methoxyethylamino)-3-nitrobenzotrifluorid

CHMPCFF1205.1EA 522 EUR
CHMPCFF1205.1
4-(2-Methoxyethylamino)-3-nitrobenzotrifluorid
4-(2-Methoxyethylamino)-3-nitrobenzotrifluorid
Formel: C₁₀H₁₁F₃N₂O₃
MDL: MFCD00798374
CAS-Nummer: 161605-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR