3-Amino-4-isopropylaminobenzotrifluorid

CHMPCFF782.EA 0 EUR
CHMPCFF782. FLUO017133.1G
3-Amino-4-isopropylaminobenzotrifluorid
3-Amino-4-isopropylaminobenzotrifluorid
Formel: C₁₀H₁₃F₃N₂
MDL: MFCD00084879
CAS-Nummer: 175277-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR