4-(Trifluormethyl)benzoesäurehydrazid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL539171-1GEA 30.8 EUR
SIAL539171-1G
4-(Trifluormethyl)benzoesäurehydrazid, Sigma-Aldrich®
4-(Trifluoromethyl)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,15 g/mol
MDL: MFCD00051703
CAS-Nummer: 339-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR