2-Hydroxy-5-methylbenzhydrazid

Lieferant: CHEMPUR

CHMPMWP01116.250EA 0 EUR
CHMPMWP01116.250
2-Hydroxy-5-methylbenzhydrazid
2-Hydroxy-5-methylbenzhydrazid
Formel: C₈H₁₀N₂O₂
CAS-Nummer: 28397-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR