Tolbutamid, Supelco®
M-Clarity™ Qualitätsstufe = MQ200
46968-250MG-REA
0
EUR
46968-250MG-R
Tolbutamid, Supelco®
Tolbutamid
Formel:
C₁₂H₁₈N₂O₃S Molecular Weight: 270,35 g/mol |
MDL:
MFCD00027169 CAS-Nummer: 64-77-7 EINECS: 200-594-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...