3-(Hydroxymethyl)phenylboronsäure

Lieferant: Apollo Scientific

APOSOR11503-1GEA 58.2 EUR
APOSOR11503-1G
3-(Hydroxymethyl)phenylboronsäure
3-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
Molecular Weight: 151,96
MDL: MFCD01317846
CAS-Nummer: 87199-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR