Pterostilben, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALP1499-10MGEA 293 EUR
SIALP1499-10MG
Pterostilben, Sigma-Aldrich®
Pterostilben
Formel: C₁₆H₁₆O₃
MDL: MFCD00238710
CAS-Nummer: 537-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR