3-Amino-4-methoxybenzolsulfonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL157031-25GEA 46.9 EUR
SIAL157031-25G
3-Amino-4-methoxybenzolsulfonsäure, Sigma-Aldrich®
3-Amino-4-methoxybenzenesulphonic acid
Formel: C₇H₉NO₄S
Molecular Weight: 203,2157 g/mol
MDL: MFCD00035759
CAS-Nummer: 98-42-0
EINECS: 202-667-5
UN: 2585
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR