2-Amino-5-methoxybenzolsulfonsäure

Lieferant: ABCR GMBH

ABCRTCA1285.0500EA 0 EUR
ABCRTCA1285.0500
2-Amino-5-methoxybenzolsulfonsäure
2-Amino-5-methoxybenzenesulphonic acid
Formel: C₇H₉NO₄S
Molecular Weight: 203,2157 g/mol
MDL: MFCD00035804
CAS-Nummer: 13244-33-2
EINECS: 236-224-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR