5-Amino-2,4,6-triiodisophthalsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL444367-50GEA 125 EUR
SIAL444367-50G
5-Amino-2,4,6-triiodisophthalsäure, Sigma-Aldrich®
5-Amino-2,4,6-triiodisophthalsäure
Formel: C₈H₄I₃NO₄
Molecular Weight: 558,84 g/mol
MDL: MFCD00190167
CAS-Nummer: 35453-19-1
EINECS: 252-575-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR