3-Iodphthalsäure
Lieferant: Thermo Scientific
MAYBBTB12976DAEA
0
EUR
MAYBBTB12976DA
3-Iodphthalsäure
3-Iodphthalsäure
Formel:
C₈H₅IO₄ Molecular Weight: 292,0300 g/mol |
MDL:
MFCD00275568 CAS-Nummer: 6937-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...