Natrium-4-aminosalicylat Dihydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL09415-50GEA
29.8
EUR
SIAL09415-50G
Natrium-4-aminosalicylat Dihydrat, Sigma-Aldrich®
Natrium-4-aminosalicylat Dihydrat
Formel:
C₇H₆NNaO₃·2H₂O Molecular Weight: 211,14 g/mol (175,12 anhy) |
MDL:
MFCD00064392 CAS-Nummer: 6018-19-5 EINECS: 205-091-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...