2-Brom-N-(3-methoxypropyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBHC-6136-5GEA
291
EUR
COBBHC-6136-5G
COBBHC-6136-1G
2-Brom-N-(3-methoxypropyl)benzolsulfonamid
2-Brom-N-(3-methoxypropyl)benzolsulfonamid
Formel:
C₁₀H₁₄BrNO₃S |
MDL:
MFCD07710755 CAS-Nummer: 848906-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...