5-Brom-2-fluorbenzamid

Lieferant: CHEMPUR

CHMPFL13777.POREA 0 EUR
CHMPFL13777.POR FLUO013777.1G
5-Brom-2-fluorbenzamid
5-Brom-2-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218,02
MDL: MFCD03094430
CAS-Nummer: 214210-17-0
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR