Potassium 3-methylthiophenyltrifluoroborate, 96%

H52638.03EA 59.6 EUR
ALFAH52638.03 ALFAH52638.MD
Potassium 3-methylthiophenyltrifluoroborate, 96%
Purity: 96%
CAS No.: 850623-48-2
Molecular Formula: C7H7BF3KS
Formula Weight: 230.10
Melting Point: 70-78°
MDL No.: MFCD04115747
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR