Potassium 2-carbamoylphenyltrifluoroborate, 96%

H52492.MDEA 72.3 EUR
ALFAH52492.MD ALFAH52492.03
Potassium 2-carbamoylphenyltrifluoroborate, 96%
Purity: 96%
CAS No.: 850623-70-0
Molecular Formula: C7H6BF3KNO
Formula Weight: 227.03
Melting Point: >300°
MDL No.: MFCD04115765
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR