3-(Trimethylsilyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL523666-5GEA 186 EUR
SIAL523666-5G
3-(Trimethylsilyl)phenylboronsäure, Sigma-Aldrich®
3-(Trimethylsilyl)phenylboronsäure
Formel: C₉H₁₅BO₂Si
MDL: MFCD03093884
CAS-Nummer: 177171-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR