3-(Trimethylsilyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8371-1GEA 92.2 EUR
COBBBB-8371-1G
3-(Trimethylsilyl)phenylboronsäure
3-(Trimethylsilyl)phenylboronsäure
Formel: C₉H₁₅BO₂Si
Molecular Weight: 194,1100 g/mol
MDL: MFCD03093884
CAS-Nummer: 177171-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR