Kaliumallyltrifluoroborat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL659274-1GEA 73.9 EUR
SIAL659274-1G
Kaliumallyltrifluoroborat, Sigma-Aldrich®
Kaliumallyltrifluoroborat
Formel: C₃H₅BF₃K
Molecular Weight: 147,976 g/mol
MDL: MFCD07368222
CAS-Nummer: 233664-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR