N,N'-Bis(salicyliden)ethylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL236071-10GEA 34.7 EUR
SIAL236071-10G
N,N'-Bis(salicyliden)ethylendiamin, Sigma-Aldrich®
N,N'-Bis(salicyliden)ethylendiamin
Formel: C₁₆H₁₆N₂O₂
Molecular Weight: 268,31 g/mol
MDL: MFCD00002244
CAS-Nummer: 94-93-9
EINECS: 202-376-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR