Acetonazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL273155-25GEA 133 EUR
SIAL273155-25G
Acetonazin, Sigma-Aldrich®
Acetonazin
Formel: C₆H₁₂N₂
MDL: MFCD00015645
CAS-Nummer: 627-70-3
EINECS: 211-009-6
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR