Diethylcyanamid, Sigma-Aldrich®

Lieferant: Merck

SIAL213810-10GEA 117 EUR
SIAL213810-10G
Diethylcyanamid, Sigma-Aldrich®
Diethylcyanamid
Formel: C₅H₁₀N₂
MDL: MFCD00001769
CAS-Nummer: 617-83-4
EINECS: 210-532-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR