Trimethylsilylacetat (Acetoxytrimethylsilan)

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS18900.25EA 52.1 EUR
SILNSILS18900.25 SILNSILS18900.100
Trimethylsilylacetat (Acetoxytrimethylsilan)
Trimethylsilylacetat (Acetoxytrimethylsilan)
Formel: C₅H₁₂O₂Si
Molecular Weight: 132,24 g/mol
MDL: MFCD00008695
CAS-Nummer: 2754-27-0
EINECS: 220-404-2
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR