3'-(N,N-Diethylamino)acetanilid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL536377-25GEA
72.8
EUR
SIAL536377-25G
3'-(N,N-Diethylamino)acetanilid, Sigma-Aldrich®
3'-(N,N-Diethylamino)acetanilid
Formel:
C₁₂H₁₈N₂O |
MDL:
MFCD00035814 CAS-Nummer: 6375-46-8 EINECS: 228-937-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...