2-Methoxyethylcyanacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL411477-25MLEA 39 EUR
SIAL411477-25ML
2-Methoxyethylcyanacetat, Sigma-Aldrich®
2-Methoxyethylcyanacetat
Formel: C₆H₉NO₃
MDL: MFCD00071550
CAS-Nummer: 10258-54-5
EINECS: 233-597-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR