2-Acetamido-4-bromanisol
Lieferant: COMBI-BLOCKS INC.
COBBHC-2834-5GEA
432
EUR
COBBHC-2834-5G
COBBHC-2834-1G
2-Acetamido-4-bromanisol
2-Acetamido-4-bromanisol
Formel:
C₉H₁₀BrNO₂ |
MDL:
MFCD12546479 CAS-Nummer: 88301-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...