Ethyl-2,4-dimethylthiazol-5-carboxylat, Sigma-Aldrich®

Lieferant: Merck

SIAL717428-1GEA 47.1 EUR
SIAL717428-1G
Ethyl-2,4-dimethylthiazol-5-carboxylat, Sigma-Aldrich®
Ethyl-2,4-dimethylthiazol-5-carboxylat
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,24 g/mol
MDL: MFCD00052874
CAS-Nummer: 7210-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR