2,4-Dimethyl-5-thiazolcarbonsäurehydrazid

Lieferant: CHEMPUR

CHMPFL9912.POREA 0 EUR
CHMPFL9912.POR FLUO009912.1G
2,4-Dimethyl-5-thiazolcarbonsäurehydrazid
2,4-Dimethyl-5-thiazolecarboxylic acid hydrazide
Formel: C₆H₉N₃OS
MDL: MFCD00832910
CAS-Nummer: 99357-25-2
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR