2,4-Dimethyl-5-thiazolcarbonsäureamid

Lieferant: CHEMPUR

CHMPFL9925.POREA 0 EUR
CHMPFL9925.POR
2,4-Dimethyl-5-thiazolcarbonsäureamid
2,4-Dimethyl-5-thiazolecarboxylic acid amide
Formel: C₆H₈N₂OS
MDL: MFCD02580045
CAS-Nummer: 66806-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR