2-Mercaptothiazol

Lieferant: ABCR GMBH

ABCRAB143996.0001EA 0 EUR
ABCRAB143996.0001
2-Mercaptothiazol
2-Mercaptothiazole
Formel: C₃H₃NS₂
Molecular Weight: 117,19 g/mol
MDL: MFCD00022449
CAS-Nummer: 82358-09-6
EINECS: 423-680-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR