2-(Methylthio)benzothiazol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL168653-50GEA
162
EUR
SIAL168653-50G
2-(Methylthio)benzothiazol, Sigma-Aldrich®
2-(Methylthio)benzothiazole
Formel:
C₈H₇NS₂ Molecular Weight: 181,2779 g/mol |
MDL:
MFCD00005784 CAS-Nummer: 615-22-5 EINECS: 210-417-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...